Learn More
Crescent Chemical Co Inc M9 MINIMAL SALTS 5X POWDER
SDP
Catalog No. 50249153
Encompass_Preferred
M9 Minimal salts 5x, Powder 52.5 g/L For cultivation of E. coli and plasmid amplification in molecular biology30 g/l Na2HPO415 g/l KH2PO45 g/l NH4Cl2.5 g/l NaClFor making 1 L 5x concentrate dissolve 52.5 g in 1 L distilled water and sterilize by autoclaving. The 5x concentra
Catalog No. 50-249-153 Supplier Crescent Chemical Co Inc Supplier No. SE48505.01
M9 Minimal salts 5x, Powder 52.5 g/L For cultivation of E. coli and plasmid amplification in molecular biology30 g/l Na2HPO415 g/l KH2PO45 g/l NH4Cl2.5 g/l NaClFor making 1 L 5x concentrate dissolve 52.5 g in 1 L distilled water and sterilize by autoclaving. The 5x concentra