Learn More
Teknova M9 Minimal Salts Agar Plates with Kanamycin-50. 100mm, 20 Plates per Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387752
Encompass
M9 minimal media is a chemically-defined growth medium that was developed for use with Escherichia coli. It is a highly versatile medium that can be used for a variety of purposes, including isolating and culturing bacteria, performing genetic transformation experiments, and growing bacteria for use in bioreactors.
Catalog No. 50-238-7752 Supplier Teknova Supplier No. M12148PK
M9 minimal media is a chemically-defined growth medium that was developed for use with Escherichia coli. It is a highly versatile medium that can be used for a variety of purposes, including isolating and culturing bacteria, performing genetic transformation experiments, and growing bacteria for use in bioreactors.