Learn More
Teknova M9 Minimal Salts Agar with 0.2% Glucose 100mm Plates. 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387760
Encompass
M9 minimal media is a chemically-defined growth medium that was developed for use with Escherichia coli. It is a highly versatile medium that can be used for a variety of purposes, including isolating and culturing bacteria, performing genetic transformation experiments, and growing bacteria for use in bioreactors.
Catalog No. 50-238-7760 Supplier Teknova Supplier No. M12604PK
M9 minimal media is a chemically-defined growth medium that was developed for use with Escherichia coli. It is a highly versatile medium that can be used for a variety of purposes, including isolating and culturing bacteria, performing genetic transformation experiments, and growing bacteria for use in bioreactors.