Learn More
Teknova M9 Minimal Salts Broth without Glucose and Ammonium Chloride, 1000mL, Sterile 8 pack.
SDP
Catalog No. 502387771
Encompass
M9 minimal media is a chemically-defined growth medium that was developed for use with Escherichia coli. It is a highly versatile medium that can be used for a variety of purposes, including isolating and culturing bacteria, performing genetic transformation experiments, and growing bacteria for use in bioreactors.
Catalog No. 50-238-7771 Supplier Teknova Supplier No. M80088PK
M9 minimal media is a chemically-defined growth medium that was developed for use with Escherichia coli. It is a highly versatile medium that can be used for a variety of purposes, including isolating and culturing bacteria, performing genetic transformation experiments, and growing bacteria for use in bioreactors.