Learn More
Sigma Aldrich Fine Chemicals Biosciences M9 Mnmal Salts 5X Lqud500ML
Catalog No. 502901616
Encompass
M9 minimal media is a highly-referenced microbial growth medium used for the cultivation of E. coli. This buffered minimal microbial medium contains only salts and nitrogen so it is traditionally supplemented with glucose amino acids and vitamins as needed.
Catalog No. 50-290-1616 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M9956500ML
M9 minimal media is a highly-referenced microbial growth medium used for the cultivation of E. coli. This buffered minimal microbial medium contains only salts and nitrogen so it is traditionally supplemented with glucose amino acids and vitamins as needed.