Learn More
Teknova M9 Salts Media with 1% Glucose and 1 mM MgSo4. 500mL Bottle, Sterile. 4 pack.
SDP
Catalog No. 502387277
Encompass
M9 minimal media is a chemically-defined growth medium that was developed for use with Escherichia coli. It is a highly versatile medium that can be used for a variety of purposes, including isolating and culturing bacteria, performing genetic transformation experiments, and growing bacteria for use in bioreactors.
Catalog No. 50-238-7277 Supplier Teknova Supplier No. 2M19904PK
M9 minimal media is a chemically-defined growth medium that was developed for use with Escherichia coli. It is a highly versatile medium that can be used for a variety of purposes, including isolating and culturing bacteria, performing genetic transformation experiments, and growing bacteria for use in bioreactors.