Learn More
Aviva Systems Biology Corp M96 Antibody - middle region (ARP34292_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34292_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against M96. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human M96. Alternate names: M96, PCL2, dJ976O13.2, RP5-976O13.1, MTF2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-52948