Learn More
Medchemexpress LLC HY-114739 10mg Medchemexpress, MA-0204 CAS:2095128-17-7 Purity:>98%
SDP
Catalog No. 501969372
Encompass_Preferred
Medchemexpress, HY-114739 10mg MA-0204 CAS:2095128-17-7 MA-0204 is a potent, highly selective and orally available peroxisome proliferator activated receptor δ (PPARδ) modulator with EC50s of 0.4 nM, 7.9 nM and 10 nM for human, mouse and rat PPARδ, respectively. Potential treatment for Duchene Muscular Dystrophy (DMD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9372 Supplier Medchemexpress LLC Supplier No. HY11473910MG
Medchemexpress, HY-114739 10mg MA-0204 CAS:2095128-17-7 MA-0204 is a potent, highly selective and orally available peroxisome proliferator activated receptor δ (PPARδ) modulator with EC50s of 0.4 nM, 7.9 nM and 10 nM for human, mouse and rat PPARδ, respectively. Potential treatment for Duchene Muscular Dystrophy (DMD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.