Learn More
Medchemexpress LLC MA-0204 25mg | 2095128-17-7 | 25 MG
SDP
Catalog No. 5000228488
Encompass_Preferred
MA-0204 is a potent, highly selective, orally available peroxisome proliferator-activated receptor delta (PPARδ) modulator supplied for research use. It is characterized by CAS 2095128-17-7, molecular weight 476.49 g/mol, and chemical formula C25H27F3N2O4. The compound is provided at high purity for in vitro and pharmacology studies; it is not for human or veterinary use.
  • Potent, selective modulation of PPARδ.
  • High purity suitable for research applications.
  • Supplied as a solid small-molecule in multiple laboratory pack sizes.
  • Appropriate for in vitro pharmacology, SAR, and preclinical studies.
  • Not for clinical, human, or veterinary use.
Catalog No. 50-002-28488 Supplier Medchemexpress LLC Supplier No. HY11473925MG
MA-0204 is a potent, highly selective, orally available peroxisome proliferator-activated receptor delta (PPARδ) modulator supplied for research use. It is characterized by CAS 2095128-17-7, molecular weight 476.49 g/mol, and chemical formula C25H27F3N2O4. The compound is provided at high purity for in vitro and pharmacology studies; it is not for human or veterinary use.
  • Potent, selective modulation of PPARδ.
  • High purity suitable for research applications.
  • Supplied as a solid small-molecule in multiple laboratory pack sizes.
  • Appropriate for in vitro pharmacology, SAR, and preclinical studies.
  • Not for clinical, human, or veterinary use.