Learn More
MEDIFY AIR LLC MA-112 REPLACEMENT FILTER SETS
SDP
Supplier:  MEDIFY AIR LLC MA112R2
Encompass_Preferred
MA-112 Replacement Filter Sets - Single. Replacement filter set includes 2 HEPA H13 Filters and 2 Activated Carbon Filters. This unit uses 2 sets of filters. All filters should be replaced after 3,000 hours of use. Air purifier comes with 2 washable pre-filters, found on the insides of the filter door panels.
Catalog No. NC1873041