Learn More
MEDIFY AIR LLC MA-14 REPLACEMENT FILTER SET
SDP
Supplier:  MEDIFY AIR LLC MA14 REPACEMENT FILTER
Encompass_Preferred
Replacement filter set includes Pre-Filter, HEPA H13 Filter, and active carbon for odor. All filters should be replaced after 2,000 hours of use.
Catalog No. NC1872926