Learn More
Medchemexpress LLC HY-107642 1mg Medchemexpress, MA-2029 CAS:287206-61-5 Purity:>98%
SDP
Catalog No. 501968447
Encompass_Preferred
Medchemexpress, HY-107642 1mg MA-2029 CAS:287206-61-5 MA-2029 is a selective, orally active, and competitive motilin receptor antagonist (IC50=4.9 nM). MA-2029 is selective for the motilin receptor over various other receptors and ion channels. MA-2029 may be useful for gastrointestinal disorders associated with disturbed gastrointestinal motility. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8447 Supplier Medchemexpress LLC Supplier No. HY1076421MG
Medchemexpress, HY-107642 1mg MA-2029 CAS:287206-61-5 MA-2029 is a selective, orally active, and competitive motilin receptor antagonist (IC50=4.9 nM). MA-2029 is selective for the motilin receptor over various other receptors and ion channels. MA-2029 may be useful for gastrointestinal disorders associated with disturbed gastrointestinal motility. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.