Learn More
MEDIFY AIR LLC MA-25 REPLACEMENT FLTR SET,2PK
SDP
Catalog No. NC1873017
Encompass
MA-25 Replacement Filter Set - Two-Pack. Replacement filter set includes 2 sets of filters: 2 Pre-Filters, 2 HEPA H13 Filters, and 2 Activated Carbon Filters. All filters should be replaced after 2500 hours of use.
Catalog No. NC1873017 Supplier MEDIFY AIR LLC Supplier No. MA25 REPACEMNT FLTR 2PCK
MA-25 Replacement Filter Set - Two-Pack. Replacement filter set includes 2 sets of filters: 2 Pre-Filters, 2 HEPA H13 Filters, and 2 Activated Carbon Filters. All filters should be replaced after 2500 hours of use.