Learn More
MEDIFY AIR LLC MA-40 REPLACEMENT FLTR SET,2PK
SDP
Catalog No. NC1873030
Encompass
MA-40 Replacement Filter Set - Two-Pack. Replacement filter set includes Pre-Filter, HEPA H13 Filter, and Activated Carbon Filter. All filters should be replaced after 3,000 hours of use.
Catalog No. NC1873030 Supplier MEDIFY AIR LLC Supplier No. MA40 REPACEMNT FLTR 2PCK
MA-40 Replacement Filter Set - Two-Pack. Replacement filter set includes Pre-Filter, HEPA H13 Filter, and Activated Carbon Filter. All filters should be replaced after 3,000 hours of use.