Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 9370MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: ADPN/1370. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b. Positive Control: HeLa cells or Kidney. Cellular Staining: Cytoplasmic ;Cell Surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human Adiponectin protein fragment. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 26-30kDa.Chromosome: 3q27. Gene Symbol: ADIPOQ. Gene ID: 9370. Ensemble: ENSG00000181092. Uniprot: NP_001171271.
Catalog No. 50-188-5337