Learn More
Medchemexpress LLC Ma-PEG4-VC-PAB-DMEA-duocarmycin DM | 99.8% | 1381.96 g/mol | C68H89ClN12O17 | 5 MG
SDP
Catalog No. 5000204379
Encompass_Preferred
MA-PEG4-VC-PAB-DMEA-duocarmycin DM is a drug-linker conjugate intended for use in the construction of antibody-drug conjugates (ADCs). The cytotoxic payload is a DNA minor-groove alkylator; the product is supplied for research use only and includes analytical documentation (COA, HNMR, RP-HPLC).
  • High purity suitable for research applications.
  • Contains a duocarmycin DM DNA minor-groove alkylator payload.
  • Solid, white to off-white appearance for easy handling.
  • Available in small research sizes for early-stage studies.
  • Analytical documentation and safety data available.
  • Recommended to store protected from light and under inert atmosphere.
Catalog No. 50-002-04379 Supplier Medchemexpress LLC Supplier No. HY1574655MG
MA-PEG4-VC-PAB-DMEA-duocarmycin DM is a drug-linker conjugate intended for use in the construction of antibody-drug conjugates (ADCs). The cytotoxic payload is a DNA minor-groove alkylator; the product is supplied for research use only and includes analytical documentation (COA, HNMR, RP-HPLC).
  • High purity suitable for research applications.
  • Contains a duocarmycin DM DNA minor-groove alkylator payload.
  • Solid, white to off-white appearance for easy handling.
  • Available in small research sizes for early-stage studies.
  • Analytical documentation and safety data available.
  • Recommended to store protected from light and under inert atmosphere.