Learn More
Meso Scale Discovery MA6000 384 Plate, MULTI-ARRAY Standard plates, 120 plates
SDP
Supplier:  Meso Scale Discovery L21XA6
Encompass
MA6000 384 PlateMSD’s uncoated MULTI-ARRAY Standard plates have a hydrophobic surface and provide a rapid, convenient method for the development of new assays and the transfer of existing immunoassays to the MSD platform.*For Research Use Only. Not for use in diagnostic procedures.
Catalog No. NC2734788
Only null left
Add to Cart
Request a Quote