Learn More
Meso Scale Discovery MA6000 384 SA Plate Pack, 30 plates
SDP
Catalog No. NC1562408
Encompass
MSD streptavidin-coated MULTI-ARRAY plates are ideal for performing sandwich immunoassays for many diverse applications using biotinylated material such as antibodies, kinase substrates, or oligonucleotides. The detection system uses patented SULFO-TAG labels that emit light upon electrochemical stimulation initiated at the electrode surfaces of the plates.
Catalog No. NC1562408 Supplier Meso Scale Discovery Supplier No. L21SA5
Add to Cart
Add to Cart
Request a Quote
MSD streptavidin-coated MULTI-ARRAY plates are ideal for performing sandwich immunoassays for many diverse applications using biotinylated material such as antibodies, kinase substrates, or oligonucleotides. The detection system uses patented SULFO-TAG labels that emit light upon electrochemical stimulation initiated at the electrode surfaces of the plates.