Learn More
Cayman Chemical Maackiain, Purity: ≥95%, CAS: 2035-15-6, 10 mg
SDP
Catalog No. 502944333
Encompass_Preferred
the histamine H1 receptor in serum-starved HeLa cells at 10, 20, or 30 µM; inhibits PMA-induced increases in the levels of mRNA encoding Il-4 in RBL-2H3 cells at 50 or 150 µM; decreases LPS-induced increases in NO production in RAW 264.7 macrophages (IC50 = 27 µM); increases survival in a mouse model of CLP-induced sepsis at 2.5 or 5 mg/kg; reduces the number of sneezes and the extent of nasal redness, swelling, and watery rhinorrhea in a rat model of TDI-induced allergies at 20 mg/kg per day
Catalog No. 50-294-4333 Supplier Cayman Chemical Supplier No. 4021510 MG
the histamine H1 receptor in serum-starved HeLa cells at 10, 20, or 30 µM; inhibits PMA-induced increases in the levels of mRNA encoding Il-4 in RBL-2H3 cells at 50 or 150 µM; decreases LPS-induced increases in NO production in RAW 264.7 macrophages (IC50 = 27 µM); increases survival in a mouse model of CLP-induced sepsis at 2.5 or 5 mg/kg; reduces the number of sneezes and the extent of nasal redness, swelling, and watery rhinorrhea in a rat model of TDI-induced allergies at 20 mg/kg per day