Learn More
Bio X Cell Monoclonal Antibody, Osteopontin 100D3, 1 MG
Supplier: Bio X Cell BE0372R001MG
The 100D3 monoclonal antibody reacts with mouse and human osteopontin (OPN), also known as SPP1. Osteopontin is a secreted arginine-glycine-aspartic acid (RGD)-containing glycoprotein found in various tissues and biological fluids. It interacts with integrins and CD44, regulating processes like bone development, immune responses, and oncogenesis.
- Clone: 100D3
- Reactivities: Mouse, Human
- Isotype: Mouse IgG2c, κ
- Reported applications: In vivo OPN neutralization, In vitro OPN neutralization, ELISA
- Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
- Endotoxin: ≤1EU/mg (≤0.001EU/μg), determined by LAL assay
- Purity: ≥95%, determined by SDS-PAGE
- Sterility: 0.2 μm filtration
- Purification method: Protein A
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C. Do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.