Learn More
Bio X Cell InVivoMAb anti-mouse osteopontin, Clone 103D6, 1 MG
Supplier: Bio X Cell BE0373R001MG
The 103D6 monoclonal antibody is an InVivoMAb anti-mouse osteopontin (SPP1) antibody. It targets mouse osteopontin (OPN), a glycoprotein that plays a role in bone development, immune responses, and oncogenesis. This blocking antibody can suppress colon tumor growth in tumor-bearing mice in vivo.
- Clone: 103D6
- Isotype: Mouse IgG2c, κ
- Target: Mouse osteopontin (SPP1)
- Applications: In vivo OPN neutralization, in vitro OPN neutralization, ELISA
- Immunogen: Recombinant mouse OPN protein
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin level: ≤1EU/mg
- Purity: ≥95% by SDS-PAGE
- Purification method: Protein G
- Molecular weight: 150 kDa
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.