Learn More
Bio X Cell mAb, Anti-Mouse Osteopontin 103D6, IgG2c, 5 MG
Supplier: Bio X Cell BE0373R005MG
This monoclonal antibody, clone 103D6, targets mouse osteopontin (OPN/SPP1), a glycoprotein involved in bone development, immune responses, and oncogenesis. It functions as a blocking antibody to neutralize OPN, enhancing tumor-specific CTL activity and suppressing tumor growth.
- Reacts with mouse osteopontin (OPN/SPP1)
- Validated for in vivo and in vitro OPN neutralization, and ELISA
- Purified by Protein G
- Formulation: PBS, pH 7.0, with no stabilizers or preservatives
- Endotoxin ≤1 EU/mg; Purity ≥95%
- Sterility: 0.2 μm filtration
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.