Learn More
Bio X Cell InVivoMAb Antibody, 103D6, Mouse Osteopontin, 50 MG
Supplier: Bio X Cell BE0373R050MG
The 103D6 monoclonal antibody reacts with mouse osteopontin (OPN), also known as SPP1. Osteopontin is a secreted arginine-glycine-aspartic acid (RGD)-containing glycoprotein that regulates diverse biological processes including bone development, immune responses, and oncogenesis.
- Immunogen: recombinant mouse OPN protein
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Sterility: 0.2 μm filtration
- Purification: protein G
- Purity: ≥95% (determined by SDS-PAGE)
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Storage: store at the stock concentration at 4°C, do not freeze
- Reported applications: in vivo OPN neutralization, in vitro OPN neutralization, ELISA
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.