Learn More
Bio X Cell Antibody, anti-mouse PD-L1, Clone 10F.2H11, 1 MG
Supplier: Bio X Cell BE0361R001MG
The 10F.2H11 monoclonal antibody reacts with mouse PD-L1 (programmed death ligand 1), also known as B7-H1 or CD274. It is a 40 kDa type I transmembrane protein that belongs to the B7 family of the Ig superfamily. This antibody has been shown to sterically and functionally block PD-L1/CD80 (B7-1) interactions, playing a role in immune response modulation.
- Isotype: rat IgG2b, κ
- Reactivity: mouse
- Clone: 10F.2H11
- Conjugation: unconjugated
- Immunogen: murine PD-L1 expressing CHO cells
- Applications: in vivo and in vitro blocking of PD-L1/CD80 (B7-1) interactions, ELISA, flow cytometry
- Formulation: PBS, pH 7.0 (no stabilizers or preservatives)
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification: protein G
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.