Learn More
Bio X Cell Anti-mouse PD-L1, 10F.2H11, Rat IgG2b, 25 MG
Supplier: Bio X Cell BE0361R025MG
The 10F.2H11 monoclonal antibody targets mouse PD-L1 (programmed death ligand 1), also known as B7-H1 or CD274. It is expressed on various immune and endothelial cells, and its binding to PD-1 inhibits TCR-mediated T cell proliferation. This antibody specifically blocks PD-L1/CD80 (B7-1) interactions.
- Unconjugated
- Immunogen: Murine PD-L1 expressing CHO cells
- Reported applications: in vivo and in vitro blocking of PD-L1/CD80 (B7-1) interactions, ELISA, flow cytometry
- Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
- Endotoxin: ≤1EU/mg (≤0.001EU/µg)
- Purity: ≥95%, determined by SDS-PAGE
- Sterility: 0.2 µm filtration
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.