Learn More
Bio X Cell Antibody, mouse PD-L1/10F.9G2, Rat IgG2b, 1 MG
Supplier: Bio X Cell BE0101R001MG
The 10F.9G2 monoclonal antibody reacts with mouse PD-L1 (programmed death ligand 1), also known as B7-H1 or CD274. This antibody blocks the interaction between PD-L1 and PD-1, and between PD-L1 and B7-1 (CD80). It is believed to play a significant role in tumor immune evasion.
- Clone: 10F.9G2
- Reactivity: mouse
- Isotype: rat IgG2b, κ
- Immunogen: mouse CD274
- Formulation: PBS, pH 6.5, no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification: protein G
- Molecular weight: 150 kDa
- Applications: in vivo PD-L1 blockade, in vitro PD-L1 blockade, immunofluorescence, immunohistochemistry (frozen), flow cytometry, western blot, in vitro organoids/organ-on-chip
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.