Learn More
Bio X Cell Antibody, anti-CA19-9, 1116-NS-19-9, Mouse IgG1, 1 MG
Supplier: Bio X Cell BE0355R001MG
The 1116-NS-19-9 monoclonal antibody targets carbohydrate antigen 19-9 (CA19-9), a tetrasaccharide overexpressed in certain cancers, such as pancreatic cancer.
- Reacts with carbohydrate antigen 19-9 (CA19-9)
- Clone: 1116-NS-19-9
- Target: carbohydrate antigen 19-9 (CA19-9)
- Host/Isotype: mouse IgG1, κ
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Production: animal-free facility, protein G purified
- Quality: ≥95% purity, ≤1 EU/mg endotoxin, 0.2 µm sterile filtered
- Applications: immunohistochemistry (paraffin), immunofluorescence, flow cytometry
- Molecular weight: 150 kDa
- Storage: store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.