Learn More
Bio X Cell anti-human CA19-9 Antibody, Mouse IgG1, 100 MG
Supplier: Bio X Cell BE0355R100MG
The InVivoMAb anti-human CA19-9 antibody, clone 1116-NS-19-9, reacts with carbohydrate antigen 19-9 (CA19-9), also known as sialylated Lewis a-antigen. CA19-9 is overexpressed in certain cancers, particularly pancreatic cancer, and is synthesized by glycosyltransferases.
- Isotype: Mouse IgG1, κ
- Reactivity: Human
- Immunogen: SW1116 human colon carcinoma cells
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/µg)
- Purity: ≥95% (determined by SDS-PAGE)
- Sterility: 0.2 µm filtration
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Applications: Immunohistochemistry (paraffin), immunofluorescence, flow cytometry
- Storage: Store at stock concentration at 4°C. Do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.