Learn More
Bio X Cell InVivoMAb anti-human CA19-9, Clone 1116-NS-19-9, 50 MG
Supplier: Bio X Cell BE0355R050MG
The 1116-NS-19-9 monoclonal antibody targets carbohydrate antigen 19-9 (CA19-9), a tetrasaccharide also known as sialylated Lewis a-antigen. CA19-9 is overexpressed in certain cancers, particularly pancreatic cancer, making this antibody suitable for research into these conditions.
- Isotype: mouse IgG1, κ
- Purified from cell culture supernatant in an animal-free facility using Protein G
- Purity: ≥95% by SDS-PAGE
- Formulated in PBS, pH 7.0, with no stabilizers or preservatives
- Applications include immunohistochemistry (paraffin), immunofluorescence, and flow cytometry
- Endotoxin: ≤1EU/mg (≤0.001EU/µg)
- Storage: store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.