Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell InVivoMAb anti-mouse CD3, Clone 145-2C11, Armenian Hamster IgG1, Unconjugated, 1 MG

Catalog No. 501528223
Encompass_Preferred

The 145-2C11 monoclonal antibody targets mouse CD3, a 20 kDa transmembrane cell-surface protein of the immunoglobulin superfamily, expressed on T lymphocytes, NK-T cells, and developing thymocytes. It plays a role in TCR signaling, T lymphocyte activation, and antigen recognition, inducing activation, proliferation, and apoptosis by binding and stimulating the TCR.

  • Isotype: Armenian Hamster IgG1
  • Recommended isotype control: Polyclonal Armenian hamster IgG
  • Recommended dilution buffer: pH 7.0 Dilution Buffer
  • Conjugation: Unconjugated (conjugation services available)
  • Immunogen: Mouse BM10-37 cytotoxic T cells
  • Reported applications: In vivo T cell depletion, in vitro T cell stimulation/activation, immunofluorescence, flow cytometry, western blot
  • Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
  • Endotoxin level: 1 EU/mg
  • Purity: 95%
  • Sterility: 0.2 m filtration
  • Production: Pu

Catalog No. 50-152-8223 Supplier Bio X Cell Supplier No. BE00011R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The 145-2C11 monoclonal antibody targets mouse CD3, a 20 kDa transmembrane cell-surface protein of the immunoglobulin superfamily, expressed on T lymphocytes, NK-T cells, and developing thymocytes. It plays a role in TCR signaling, T lymphocyte activation, and antigen recognition, inducing activation, proliferation, and apoptosis by binding and stimulating the TCR.

  • Isotype: Armenian Hamster IgG1
  • Recommended isotype control: Polyclonal Armenian hamster IgG
  • Recommended dilution buffer: pH 7.0 Dilution Buffer
  • Conjugation: Unconjugated (conjugation services available)
  • Immunogen: Mouse BM10-37 cytotoxic T cells
  • Reported applications: In vivo T cell depletion, in vitro T cell stimulation/activation, immunofluorescence, flow cytometry, western blot
  • Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
  • Endotoxin level: 1 EU/mg
  • Purity: 95%
  • Sterility: 0.2 m filtration
  • Production: Pu

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.