Learn More
Bio X Cell Antibody, Human/Mouse Ganglioside GD2, 14G2a, 5 MG
Supplier: Bio X Cell BE0318R005MG
The 14G2a monoclonal antibody reacts with human ganglioside GD2, a sialic-acid bearing glycolipid involved in mediating cell attachment to the extracellular matrix. Ganglioside GD2 is expressed on tumors of neuroectodermal origin, such as human neuroblastoma and melanoma, making it a target for immunotherapy with monoclonal antibodies or artificial T cell receptors.
- Clone: 14G2a
- Reactivities: Mouse, Human
- Isotype: Mouse IgG2a, κ
- Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.