Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Anti-Mouse CD3 Antibody, Clone 17A2, 1 MG

Catalog No. 501528226
Encompass_Preferred

The 17A2 monoclonal antibody reacts with mouse CD3, a transmembrane cell-surface protein involved in TCR signaling, T lymphocyte activation, and antigen recognition.

  • Isotype: Rat IgG2b, κ
  • Immunogen: γδ TCR-positive T-T hybridoma D1
  • Purity: ≥95% (SDS-PAGE)
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg)
  • Sterility: 0.2 μm filtered
  • Purification: Protein G, animal-free facility
  • Formulation: PBS, pH 7.0, no stabilizers or preservatives
  • Storage: Store at 4°C, do not freeze
  • Applications: In vitro T cell stimulation/activation
  • Molecular weight: 150 kDa

Catalog No. 50-152-8226 Supplier Bio X Cell Supplier No. BE0002R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The 17A2 monoclonal antibody reacts with mouse CD3, a transmembrane cell-surface protein involved in TCR signaling, T lymphocyte activation, and antigen recognition.

  • Isotype: Rat IgG2b, κ
  • Immunogen: γδ TCR-positive T-T hybridoma D1
  • Purity: ≥95% (SDS-PAGE)
  • Endotoxin: ≤1EU/mg (≤0.001EU/μg)
  • Sterility: 0.2 μm filtered
  • Purification: Protein G, animal-free facility
  • Formulation: PBS, pH 7.0, no stabilizers or preservatives
  • Storage: Store at 4°C, do not freeze
  • Applications: In vitro T cell stimulation/activation
  • Molecular weight: 150 kDa

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.