Learn More
Bio X Cell Monoclonal Antibody, anti-mouse Thy1.1 19E12, 1 MG
Supplier: Bio X Cell BE0214R001MG
The 19E12 monoclonal antibody reacts with mouse Thy1.1 (CD90.1), a 25-35 kDa GPI-anchored protein expressed by thymocytes, peripheral T cells, myoblasts, epidermal cells, and keratinocytes. This antibody is particularly useful for depletion of T lymphocytes.
- Clone: 19E12
- Isotype: Mouse IgG2a, κ
- Immunogen: AKR mouse SL3 leukemia cells
- Reported applications: In vivo T cell depletion
- Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtered
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.