Learn More
Bio X Cell Monoclonal Antibody, mouse IL-10R, 1B1.3A, 1 MG
Supplier: Bio X Cell BE0050R001MG
The 1B1.3A monoclonal antibody reacts with mouse IL-10R (CD210), a class II cytokine receptor. It mediates immunoregulation and inflammation by downregulating pro-inflammatory cytokines and enhancing B lymphocyte functions.
- Neutralizing antibody; blocks human IL-10 binding to mouse IL-10R
- Does not recognize human IL-10R
- Isotype: Rat IgG1, κ
- Applications: In vivo/in vitro IL-10/IL-10R signaling blockade, Flow cytometry, Western Blot
- Formulation: PBS, pH 6.5, 0.01% Tween, preservative-free
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.