Learn More
Bio X Cell mouse IL-10R 1B1.3A, Rat IgG1, 25 MG
Supplier: Bio X Cell BE0050R025MG
The 1B1.3A monoclonal antibody reacts with mouse IL-10R (CD210), a class II cytokine receptor expressed on various immune cells. It is a neutralizing antibody that blocks human IL-10 binding to mouse IL-10R, playing a role in immunoregulation and inflammation.
- Reacts with mouse IL-10R (CD210).
- Neutralizing antibody; blocks human IL-10 binding to mouse IL-10R.
- Formulated in PBS, pH 6.5, 0.01% Tween.
- Endotoxin level ≤1 EU/mg (≤0.001 EU/μg).
- Purity ≥95% by SDS-PAGE.
- Sterilized by 0.2 μm filtration.
- Purified by Protein G.
- Store at 4°C; do not freeze.
- Applications: in vivo/in vitro blocking, flow cytometry, western blot.
- Recombinant murine chimeric versions available.
- Fc-silenced versions available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.