Learn More
Bio X Cell Bio X Cell, anti-human NKG2D (CD314), Clone 1D11, 100 MG
Supplier: Bio X Cell BE0351R100MG
The 1D11 monoclonal antibody reacts with human NKG2D (CD314), a type II transmembrane glycoprotein. It is expressed on NK cells, NKT cells, CD8 T cells, γ/δ T cells, and macrophages, and has been implicated in anti-tumor surveillance and immune responses.
- Isotype: Mouse IgG1, κ
- Immunogen: Human NKL cells
- Applications: In vitro NKG2D blocking, immunoprecipitation, flow cytometry
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtered
- Production: Purified from animal-free cell culture supernatant
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.