Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell InVivoMAb anti-TGF-beta, 1D11.16.8, Mouse IgG1, 1 MG

Catalog No. 501528284
Encompass_Preferred

The 1D11.16.8 monoclonal antibody reacts with mouse, human, rat, monkey, hamster, canine, and bovine TGF-β (transforming growth factor beta) isoforms 1, 2, and 3. It is a neutralizing antibody targeting a multifunctional cytokine involved in cell proliferation, immune function, tissue remodeling, and wound repair.

  • Clone: 1D11.16.8
  • Target: TGF-β isoforms 1, 2, and 3
  • Host: Mouse
  • Isotype: Mouse IgG1, κ
  • Unconjugated
  • Applications: In vivo TGFβ neutralization, in vitro TGFβ neutralization, Western blot
  • Formulation: PBS, pH 7.0; no stabilizers or preservatives
  • Purity: ≥95% by SDS-PAGE
  • Endotoxin: ≤1EU/mg
  • Sterility: 0.2 µm filtration
  • Production: Animal-free purified
  • Purification: Protein G
  • Molecular Weight: 150 kDa
  • Storage: 4°C, do not freeze

Catalog No. 50-152-8284 Supplier Bio X Cell Supplier No. BE0057R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The 1D11.16.8 monoclonal antibody reacts with mouse, human, rat, monkey, hamster, canine, and bovine TGF-β (transforming growth factor beta) isoforms 1, 2, and 3. It is a neutralizing antibody targeting a multifunctional cytokine involved in cell proliferation, immune function, tissue remodeling, and wound repair.

  • Clone: 1D11.16.8
  • Target: TGF-β isoforms 1, 2, and 3
  • Host: Mouse
  • Isotype: Mouse IgG1, κ
  • Unconjugated
  • Applications: In vivo TGFβ neutralization, in vitro TGFβ neutralization, Western blot
  • Formulation: PBS, pH 7.0; no stabilizers or preservatives
  • Purity: ≥95% by SDS-PAGE
  • Endotoxin: ≤1EU/mg
  • Sterility: 0.2 µm filtration
  • Production: Animal-free purified
  • Purification: Protein G
  • Molecular Weight: 150 kDa
  • Storage: 4°C, do not freeze

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.