Learn More
Bio X Cell Antibody, anti-human NKG2D, Clone 1D11, 5 MG
Supplier: Bio X Cell BE0351R005MG
The 1D11 monoclonal antibody reacts with human NKG2D (CD314), a type II transmembrane lectin-like glycoprotein. Expressed on NK cells, NKT cells, CD8 T cells, γ/δ T cells, and macrophages, NKG2D is implicated in anti-tumor surveillance, immune response against viral infection, and diabetes progression. Studies show 1D11 blocks NKG2D binding to ligands or stimulates NKG2D-positive cells.
- Isotype: mouse IgG1, κ
- Immunogen: human NKL cells
- Applications: in vitro NKG2D blocking, immunoprecipitation, flow cytometry
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg)
- Purity: ≥95% by SDS-PAGE
- Production & purification: animal-free, protein G
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.