Learn More
Bio X Cell BioXCell Mouse CD19 1D3 Rat IgG2a Unconjugated, 100 MG
Supplier: Bio X Cell BE0150R100MG
This 1D3 monoclonal antibody reacts with mouse CD19, a B cell-specific 95 kDa transmembrane glycoprotein. CD19 is highly expressed in most lymphomas and leukemias, making it an attractive target for immunotherapy. This antibody functions as a positive regulator of B-cell receptor signaling and has been reported to deplete B cells, often requiring combination treatment for optimal effect.
- Endotoxin levels ≤1EU/mg (≤0.001EU/μg)
- Purity ≥95% determined by SDS-PAGE
- Sterility via 0.2 μm filtration
- Produced in an animal-free facility
- Purified using Protein G
- Molecular weight of 150 kDa
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.