Learn More
Bio X Cell Antibody, Human CSF1R, Clone 2-4A5-4, 100 MG
Supplier: Bio X Cell BE0347R100MG
The 2-4A5-4 monoclonal antibody reacts with human colony stimulating factor 1 receptor (CSF1R), also known as CD115. CSF1R is a single-pass type I membrane protein expressed on monocytes/macrophages and osteoclasts, regulating proliferation and differentiation in the monocytic lineage.
- Purified from cell culture supernatant in an animal-free facility using Protein G.
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives.
- Endotoxin level: ≤1EU/mg (≤0.001EU/μg).
- Purity: ≥95% (determined by SDS-PAGE).
- Sterility: 0.2 μm filtration.
- Reported applications include in vitro CSF1R neutralization, immunohistochemistry (paraffin), functional assays, and flow cytometry.
- Storage: store at stock concentration at 4°C, do not freeze.
- Molecular weight: 150 kDa.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.