Learn More
Bio X Cell Antibody, anti-human CSF1R, Clone 2-4A5-4, 25 MG
Supplier: Bio X Cell BE0347R025MG
This 2-4A5-4 monoclonal antibody reacts with human colony stimulating factor 1 receptor (CSF1R/CD115). CSF1R is expressed by monocytes/macrophages, dendritic cells, and osteoclasts, regulating proliferation and differentiation in the monocytic lineage.
- Clone: 2-4A5-4
- Reactivities: Human
- Isotype: Rat IgG1, κ
- Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C; do not freeze
- Reported applications: In vitro CSF1R neutralization, immunohistochemistry (paraffin), functional assays, flow cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.