Learn More
Bio X Cell Antibody, CSF1R (CD115), Clone 2-4A5-4, 50 MG
Supplier: Bio X Cell BE0347R050MG
The 2-4A5-4 monoclonal antibody targets human colony stimulating factor 1 receptor (CSF1R), also known as CD115. CSF1R is a single-pass type I membrane protein that regulates the proliferation and differentiation of cells in the monocytic lineage.
- Clone: 2-4A5-4
- Reactivity: Human
- Isotype: Rat IgG1, κ
- Unconjugated
- Immunogen: Rat NRK cells expressing transduced human CSF1R
- Formulation: PBS, pH 7.0 (no stabilizers or preservatives)
- Endotoxin: ≤1 EU/mg
- Purity: ≥95% (SDS-PAGE)
- Sterility: 0.2 μm filtration
- Purified from cell culture supernatant
- Purification method: Protein G
- Molecular weight: 150 kDa
- Applications: In vitro CSF1R neutralization, immunohistochemistry (paraffin), functional assays, flow cytometry
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.