Learn More
Bio X Cell InVivoMAb Anti-Mouse CD16/CD32, Clone 2.4G2, 100 MG
Supplier: Bio X Cell BE0307R100MG
The InVivoMAb anti-mouse CD16/CD32 monoclonal antibody (Clone 2.4G2) specifically reacts with mouse CD16 (FcγRIII) and CD32 (FcγRII), preventing non-specific binding of the Fc portion of IgG. It is suitable for in vivo Fc receptor blocking, flow cytometry, and immunofluorescence applications.
- Purified from cell culture supernatant in an animal-free facility
- Protein G purification method
- Purity ≥95%, determined by SDS-PAGE
- Sterility achieved via 0.2 μm filtration
- Formulated in PBS, pH 8.0, without stabilizers or preservatives
- Endotoxin level ≤1EU/mg
- Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.