Learn More
Bio X Cell Antibody, Clone 2.4G2, Mouse CD16/CD32, 50 MG
Supplier: Bio X Cell BE0307R050MG
This 2.4G2 monoclonal antibody targets mouse CD16 (FcγRIII) and CD32 (FcγRII) receptors, which are found on various immune cells and bind to the Fc portion of antibody-antigen complexes. It is utilized in experiments such as flow cytometry and immunofluorescence to prevent non-specific Fc binding.
- Clone 2.4G2, Rat IgG2b, κ isotype
- Specifically reacts with mouse CD16 (FcγRIII) and CD32 (FcγRII)
- Applications include in vivo Fc receptor blocking, flow cytometry, and immunofluorescence
- Immunogen: BALB/c mouse macrophage cell line J774
- Formulation: PBS, pH 8.0, no stabilizers or preservatives
- Purity: ≥95% by SDS-PAGE; Endotoxin: ≤1 EU/mg
- Purified by Protein G; Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Storage at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.