Learn More
Bio X Cell Clone 20D5, Rat IgG2a, Mouse, 1 MG
Supplier: Bio X Cell BE0321R001MG
The 20D5 monoclonal antibody reacts with mouse NKG2A, NKG2C, and NKG2E receptors. It forms heterodimers with CD94, primarily expressed on NK and NKT cells, and activated CD8+ T cells. This antibody helps promote anti-tumor immunity by blocking NKG2A signaling, making it valuable for research into immune checkpoint inhibitory therapies.
- Reacts with mouse NKG2A, NKG2C, and NKG2E
- Isotype: rat IgG2a, κ
- Unconjugated for flexible use
- Validated for in vivo and in vitro NKG2A blockade, immunohistochemistry (frozen), and flow cytometry
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Purity: ≥95% with ≤1EU/mg endotoxin
- Purified from cell culture supernatant in an animal-free facility
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.