Learn More
Bio X Cell InVivoMAb Antibody, anti-mouse NKG2A/C/E, Clone 20D5, Rat IgG2a, Unconjugated, 100 MG
Supplier: Bio X Cell BE0321R100MG
The 20D5 monoclonal antibody targets mouse NKG2A, NKG2C, and NKG2E receptors, which form heterodimers with CD94. These complexes are primarily found on NK, NKT, and activated CD8+ T cells, recognizing Qa-1. NKG2A transmits inhibitory signals, while NKG2C and NKG2E transmit stimulatory signals. Blocking NKG2A signaling enhances anti-tumor immunity in murine models by boosting T and NK cell activity, making NKG2A a potential target for immune checkpoint therapy.
- Targets mouse NKG2A, NKG2C, and NKG2E
- Clone 20D5
- Isotype: Rat IgG2a,
- Unconjugated
- Formulated in PBS, pH 7.0 (no stabilizers or preservatives)
- Endotoxin level: 1 EU/mg
- Purity: 95%
- Sterilized by 0.2 m filtration
- Animal-free production
- Purified via Protein G
- Store at 4C, do not freeze
- Reported applications include in vivo and in vitro NKG2A blockade, immunohistochemistry (frozen), and flow cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.