Learn More
Bio X Cell InVivoMAb, NKG2A/C/E, 20D5, Rat IgG2a, 50 MG
Supplier: Bio X Cell BE0321R050MG
The 20D5 monoclonal antibody reacts with mouse NKG2A, NKG2C, and NKG2E (CD159a, CD159c, and CD159e). These NKG2 receptors are C-type lectin-like receptors that form heterodimers with CD94 and are primarily expressed on NK cells and NKT cells. Blocking NKG2A signaling has been shown to promote anti-tumor immunity in murine tumor models, making NKG2A a target for novel immune checkpoint inhibitory therapy.
- Host species: Rat IgG2a, κ
- Target species: Mouse NKG2A, NKG2C, and NKG2E
- Applications: In vivo NKG2A blockade, in vitro NKG2A blockade, IHC (frozen), flow cytometry
- Formulation: PBS, pH 7.0 (no stabilizers/preservatives)
- Endotoxin: ≤1EU/mg
- Purity: ≥95% (SDS-PAGE)
- Sterility: 0.2 μm filtered
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.