Learn More
Bio X Cell InVivoMAb anti-mouse PD-1, Rat IgG2a, 1 MG
Supplier: Bio X Cell BE0273R001MG
InVivoMAb anti-mouse PD-1 (CD279) is an antibody solution specifically designed for immunological research targeting mouse PD-1. This product is suitable for a wide range of in vivo and in vitro applications.
- In vivo blocking of PD-1/PD-L signaling
- In vitro PD-1 neutralization
- Immunohistochemistry (frozen)
- Immunofluorescence
- Western blot
- Flow cytometry
- Antibody host: Rat
- Species reactivity: Mouse
- Antibody isotype: IgG2a
- Store at stock concentration at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.