Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio X Cell Monoclonal Antibody, Mouse CD28, IgG2, 1 MG

Catalog No. 501528241
Encompass_Preferred

The InVivoMAb anti-mouse CD28 antibody is designed for various research applications involving mouse CD28, including in vitro T cell stimulation/activation and in vivo CD28 blockade studies.

  • Applications: in vitro T cell stimulation/activation, in vivo CD28 blockade
  • Antibody host: Syrian Hamster
  • Species reactivity: Mouse
  • Antibody isotype: IgG2
  • Immunogen: C57BL/6 mouse T cell lymphoma EL-4 cells
  • Storage temperature: 4°C, do not freeze

Catalog No. 50-152-8241 Supplier Bio X Cell Supplier No. BE00151R001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The InVivoMAb anti-mouse CD28 antibody is designed for various research applications involving mouse CD28, including in vitro T cell stimulation/activation and in vivo CD28 blockade studies.

  • Applications: in vitro T cell stimulation/activation, in vivo CD28 blockade
  • Antibody host: Syrian Hamster
  • Species reactivity: Mouse
  • Antibody isotype: IgG2
  • Immunogen: C57BL/6 mouse T cell lymphoma EL-4 cells
  • Storage temperature: 4°C, do not freeze

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.