Learn More
Bio X Cell Antibody, TIM-1 (CD365), Clone 3D10, Rat IgG1, 1 MG
Supplier: Bio X Cell BE0314R001MG
This monoclonal antibody, clone 3D10, targets mouse T cell immunoglobulin and mucin domain 1 (TIM-1), also known as CD365. TIM-1, a type I cell-surface glycoprotein within the Ig superfamily, is predominantly expressed on TH2 cells and acts as a stimulatory molecule for T cell activation. It plays crucial roles in regulating immune responses to viral infections, allergic reactions, asthma, and transplant tolerance. This antibody has been shown to block TIM-1 in vivo and enhance atherosclerosis in mice studies.
- Reacts with mouse TIM-1 (CD365)
- Isotype: Rat IgG1,
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Low endotoxin level (1EU/mg)
- Purity greater than 95%
- Sterility achieved by 0.2 m filtration
- Suitable for in vivo and in vitro TIM-1 blockade
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.